Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252600.1 | NP_001239529.1 | 4 - 5 | 838 | 67 | |
NM_001252601.1 | NP_001239530.1 | 4 - 5 | 838 | 67 | ||
NM_016850.3 | NP_058546.1 | 4 - 5 | 838 | 67 | ||
GenBank mRNA | AK002830.1 | - | 4 - 5 | 66 | ||
AK004349.1 | - | 4 - 5 | 66 | |||
AK079685.1 | - | 4 - 5 | 838 | 67 | ||
AK089869.1 | - | 4 - 5 | 838 | 67 | ||
AK150403.1 | - | 4 - 5 | 838 | 67 | ||
AK150608.1 | - | 4 - 5 | 838 | 67 | ||
AK151470.1 | - | 4 - 5 | 838 | 67 | ||
AK152401.1 | - | 4 - 5 | 838 | 67 | ||
AK152403.1 | - | 4 - 5 | 838 | 67 | ||
AK153054.1 | - | 4 - 5 | 838 | 67 | ||
AK158378.1 | - | 4 - 5 | 837 | 67 | ||
AK158403.1 | - | 4 - 5 | 837 | 67 | ||
AK159883.1 | - | 4 - 5 | 838 | 67 | ||
BC138799.1 | - | 3 - 4 | 778 | 8861821 | 67 | |
DT923384.1 | - | 2 - 3 | 315 | 67 | ||
U73037.1 | - | 3 - 4 | 815 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |