Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170982.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170982.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170984.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_001170984.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_016884.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
NM_016884.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015162.mm.1 |
TC1400002023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170982.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170982.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170984.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_001170984.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_016884.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
NM_016884.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015160.mm.1 |
TC1400002023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015161.mm.1 |
NM_016884.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015161.mm.1 |
NM_016884.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015161.mm.1 |
TC1400002023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170982.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170982.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170984.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_001170984.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_016884.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
NM_016884.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015159.mm.1 |
TC1400002023.mm.1 |
MTA TC |