Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018782.2 | NP_061252.2 | 6 - 7 | 698 | 59 | |
GenBank mRNA | AB015595.1 | - | 6 - 7 | 702 | 59 | |
AF146525.1 | - | 4 - 5 | 375 | 59 | ||
AF209905.1 | - | 2 - 3 | 194 | 59 | ||
AK137210.1 | - | 6 - 7 | 697 | 59 | ||
BC138537.1 | - | 3 - 4 | 325 | 8861557 | 59 | |
BC138540.1 | - | 3 - 4 | 325 | 8861560 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |