Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018869.3 | NP_061357.3 | 5 - 6 | 1268 | 64 | |
GenBank mRNA | AF040746.1 | - | 5 - 6 | 678 | 64 | |
AK135076.1 | - | 5 - 6 | 1270 | 64 | ||
AK161821.1 | - | 5 - 6 | 748 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |