Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018883.2 | NP_061371.2 | 14 - 15 | 1501 | 72 | |
GenBank mRNA | AF117384.1 | - | 14 - 15 | 1456 | 72 | |
AK144483.1 | - | 13 - 14 | 1771 | 72 | ||
BC017529.1 | - | 14 - 15 | 1466 | 4924656 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |