Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145979.1 | NP_001139451.1 | 7 - 8 | 1171 | 96 | |
NM_019581.3 | NP_062527.2 | 7 - 8 | 1206 | 96 | ||
GenBank mRNA | AB024573.1 | - | 5 - 6 | 842 | 96 | |
AF168991.1 | - | 7 - 8 | 1111 | 96 | ||
AK087511.1 | - | 7 - 8 | 1077 | 96 | ||
AK146415.1 | - | 7 - 8 | 1156 | 96 | ||
BC049089.1 | - | 7 - 8 | 1167 | 5720710 | 96 | |
BC051958.1 | - | 7 - 8 | 1167 | 5720710 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |