Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019649.2 | NP_062623.2 | 7 - 8 | 814 | 65 | |
GenBank mRNA | AK152410.1 | - | 7 - 8 | 804 | 65 | |
AK152809.1 | - | 7 - 8 | 804 | 65 | ||
AK165742.1 | - | 6 - 7 | 1508 | 65 | ||
AK171901.1 | - | 7 - 8 | 805 | 65 | ||
BC022172.1 | - | 7 - 8 | 805 | 4193279 | 65 | |
D67067.1 | - | 7 - 8 | 801 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |