Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019795.4 | NP_062769.2 | 3 - 4 | 858 | 83 | |
GenBank mRNA | AB028861.1 | - | 3 - 4 | 306 | 83 | |
AK076032.1 | - | 3 - 4 | 329 | 83 | ||
AK145506.1 | - | 3 - 4 | 320 | 83 | ||
AK145739.1 | - | 3 - 4 | 320 | 83 | ||
AK166872.1 | - | 3 - 4 | 320 | 83 | ||
AY323828.1 | - | 3 - 4 | 294 | 83 | ||
BC023681.1 | - | 3 - 4 | 502 | 5343384 | 83 | |
BC055729.1 | - | 3 - 4 | 857 | 6414499 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |