Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hacl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019975.3 | NP_064359.2 | 13 - 14 | 1346 | 97 | |
GenBank mRNA | AJ132139.1 | - | 13 - 14 | 1344 | 97 | |
AK005505.1 | - | 13 - 14 | 1333 | 97 | ||
AK041686.1 | - | 13 - 14 | 1342 | 97 | ||
AK050078.1 | - | 13 - 14 | 1346 | 97 | ||
AK161488.1 | - | 1326 | 97 | |||
BC021360.1 | - | 13 - 14 | 1277 | 5011451 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |