Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038010.2 | NP_001033099.1 | 9 - 10 | 1472 | 71 | |
NM_020004.5 | NP_064388.2 | 9 - 10 | 1475 | 71 | ||
GenBank mRNA | AF254441.1 | - | 9 - 10 | 1412 | 71 | |
AK137358.1 | - | 9 - 10 | 1452 | 71 | ||
AK145205.1 | - | 7 - 8 | 1472 | 71 | ||
AK147042.1 | - | 9 - 10 | 1420 | 71 | ||
AK150225.1 | - | 9 - 10 | 1453 | 71 | ||
AK154866.1 | - | 9 - 10 | 1463 | 71 | ||
AK155347.1 | - | 9 - 10 | 1649 | 71 | ||
AK155791.1 | - | 9 - 10 | 1474 | 71 | ||
AK155842.1 | - | 9 - 10 | 1473 | 71 | ||
AK158079.1 | - | 9 - 10 | 1463 | 71 | ||
BC011088.1 | - | 9 - 10 | 1382 | 71 | ||
BC051952.1 | - | 9 - 10 | 1457 | 71 | ||
BC060050.1 | - | 9 - 10 | 1457 | 71 | ||
BC063752.1 | - | 9 - 10 | 1457 | 6333090 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |