Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020603.2 | NP_065628.1 | 8 - 9 | 886 | 66 | |
GenBank mRNA | AK145068.1 | - | 8 - 9 | 941 | 66 | |
BC013527.1 | - | 8 - 9 | 929 | 66 | ||
BC026431.1 | - | 5 - 6 | 510 | 66 | ||
BC046977.1 | - | 8 - 9 | 886 | 6315172 | 66 | |
KJ650211.1 | - | 8 - 9 | 871 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |