Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020605.3 | NP_065630.1 | 2 - 3 | 1220 | 61 | |
GenBank mRNA | AB024449.1 | - | 2 - 3 | 1216 | 61 | |
AK045706.1 | - | 2 - 3 | 1207 | 61 | ||
AK046136.1 | - | 1213 | 61 | |||
AK082880.1 | - | 2 - 3 | 1209 | 61 | ||
BC103682.1 | - | 2 - 3 | 1175 | 40047806 | 61 | |
BC104736.1 | - | 2 - 3 | 1175 | 40047803 | 61 | |
BC105307.1 | - | 2 - 3 | 1175 | 40047801 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |