Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021328.3 | NP_067303.1 | 4 - 5 | 201 | 80 | |
GenBank mRNA | AF244361.1 | - | 4 - 5 | 181 | 80 | |
AF271733.1 | - | 4 - 5 | 244 | 80 | ||
AK005992.1 | - | 4 - 5 | 210 | 80 | ||
AK040685.1 | - | 4 - 5 | 201 | 80 | ||
AK170915.1 | - | 4 - 5 | 242 | 80 | ||
BC026543.1 | - | 4 - 5 | 186 | 5040094 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |