Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021449.3 | NP_067424.2 | 4 - 5 | 563 | 71 | |
NM_175357.3 | NP_780566.1 | 4 - 5 | 566 | 71 | ||
GenBank mRNA | AF229032.1 | - | 4 - 5 | 518 | 71 | |
AK032836.1 | - | 4 - 5 | 540 | 71 | ||
AK050557.1 | - | 4 - 5 | 549 | 71 | ||
AK075959.1 | - | 4 - 5 | 540 | 71 | ||
AK076144.1 | - | 4 - 5 | 546 | 71 | ||
AK077707.1 | - | 4 - 5 | 546 | 71 | ||
AK141428.1 | - | 4 - 5 | 541 | 71 | ||
AK160219.1 | - | 4 - 5 | 542 | 71 | ||
BC033458.1 | - | 4 - 5 | 524 | 71 | ||
BC046967.1 | - | 3 - 4 | 481 | 71 | ||
BC069905.1 | - | 4 - 5 | 523 | 71 | ||
BC086488.1 | - | 4 - 5 | 541 | 30443656 | 71 | |
BQ749260.1 | - | 4 - 5 | 511 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |