Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012284.mm.1 |
TC0500001390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012284.mm.1 |
NM_024216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012284.mm.1 |
NM_024216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012286.mm.1 |
TC0500001390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012286.mm.1 |
NM_024216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012286.mm.1 |
NM_024216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012288.mm.1 |
TC0500001390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012288.mm.1 |
NM_024216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012288.mm.1 |
NM_024216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012285.mm.1 |
TC0500001390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012285.mm.1 |
NM_024216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012285.mm.1 |
NM_024216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012287.mm.1 |
TC0500001390.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012287.mm.1 |
NM_024216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012287.mm.1 |
NM_024216.1 |
Refseq |