Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024223.2 | NP_077185.1 | 3 - 4 | 672 | 78 | |
GenBank mRNA | AF469648.1 | - | 3 - 4 | 378 | 78 | |
AK002484.1 | - | 3 - 4 | 357 | 78 | ||
BC002093.1 | - | 3 - 4 | 375 | 3489018 | 78 | |
BC002096.1 | - | 3 - 4 | 358 | 3493903 | 78 | |
BC007149.1 | - | 3 - 4 | 358 | 78 | ||
CT010309.1 | - | 3 - 4 | 336 | 78 | ||
CX217539.1 | - | 3 - 4 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |