Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025727.3 | NP_080003.1 | 1 - 2 | 296 | 69 | |
GenBank mRNA | AK006288.1 | - | 1 - 2 | 283 | 69 | |
AK014906.1 | - | 1 - 2 | 297 | 69 | ||
AK077202.1 | - | 1 - 2 | 298 | 69 | ||
AY495337.1 | - | 1 - 2 | 297 | 69 | ||
BC064049.1 | - | 1 - 2 | 293 | 69 | ||
BC139267.1 | - | 1 - 2 | 272 | 8862289 | 69 | |
BC139268.1 | - | 1 - 2 | 272 | 8862290 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |