Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025903.2 | NP_080179.1 | 7 - 8 | 944 | 77 | |
GenBank mRNA | AK002702.1 | - | 1 - 2 | 169 | 77 | |
AK007687.1 | - | 7 - 8 | 945 | 77 | ||
AK149496.1 | - | 7 - 8 | 925 | 77 | ||
AK156625.1 | - | 6 - 7 | 1020 | 77 | ||
AK165967.1 | - | 7 - 8 | 938 | 77 | ||
AK166130.1 | - | 7 - 8 | 1012 | 77 | ||
AK166258.1 | - | 7 - 8 | 902 | 77 | ||
AY298860.1 | - | 7 - 8 | 773 | 77 | ||
BC023486.1 | - | 7 - 8 | 891 | 77 | ||
BC037434.1 | - | 7 - 8 | 900 | 2651172 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |