Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026121.3 | NP_080397.1 | 2 - 3 | 413 | 76 | |
GenBank mRNA | AF332863.1 | - | 2 - 3 | 406 | 76 | |
AK010765.1 | - | 2 - 3 | 404 | 76 | ||
AK136899.1 | - | 2 - 3 | 403 | 76 | ||
AK160316.1 | - | 2 - 3 | 405 | 76 | ||
AK162658.1 | - | 2 - 3 | 400 | 76 | ||
BC037239.1 | - | 2 - 3 | 411 | 76 | ||
BC058518.1 | - | 2 - 3 | 396 | 5717401 | 76 | |
BC138201.1 | - | 2 - 3 | 491 | 8861220 | 76 | |
BC145986.1 | - | 2 - 3 | 491 | 40131289 | 76 | |
BU939693.1 | - | 418 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |