Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026496.4 | NP_080772.2 | 5 - 6 | 1083 | 70 | |
GenBank mRNA | AF411213.1 | - | 5 - 6 | 816 | 70 | |
AK002633.1 | - | 5 - 6 | 1084 | 70 | ||
BC055035.1 | - | 5 - 6 | 872 | 6306106 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |