Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026530.5 | NP_080806.4 | 10 - 11 | 1245 | 82 | |
GenBank mRNA | AK012128.1 | - | 9 - 10 | 1200 | 82 | |
AK146973.1 | - | 10 - 11 | 1230 | 82 | ||
AK160356.1 | - | 10 - 11 | 1214 | 82 | ||
AK172381.1 | - | 9 - 10 | 988 | 82 | ||
BC017625.1 | - | 9 - 10 | 1194 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |