Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026530.5 | NP_080806.4 | 11 - 12 | 1338 | 63 | |
GenBank mRNA | AK012128.1 | - | 10 - 11 | 1293 | 63 | |
AK146973.1 | - | 11 - 12 | 1323 | 63 | ||
AK160356.1 | - | 11 - 12 | 1307 | 63 | ||
AK172381.1 | - | 10 - 11 | 1081 | 63 | ||
BC017625.1 | - | 10 - 11 | 1287 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |