Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027027.2 | NP_081303.1 | 324 | 67 | ||
GenBank mRNA | AF398970.1 | - | 2 - 3 | 256 | 67 | |
AK005878.1 | - | 325 | 67 | |||
BC061038.1 | - | 316 | 6703903 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |