Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027897.4 | NP_082173.3 | 5 - 6 | 573 | 56 | |
GenBank mRNA | AK004849.1 | - | 5 - 6 | 496 | 56 | |
AK050214.1 | - | 5 - 6 | 487 | 56 | ||
BC100314.1 | - | 5 - 6 | 480 | 30848276 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |