Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB047647.1 | - | 1 - 2 | 184 | 69 | |
AB052779.1 | - | 3 - 4 | 441 | 69 | ||
AB182651.1 | - | 3 - 4 | 559 | 69 | ||
AB182652.1 | - | 1 - 2 | 184 | 69 | ||
AF287260.1 | - | 1 - 2 | 322 | 69 | ||
AK005918.1 | - | 1 - 2 | 341 | 69 | ||
AK019928.1 | - | 3 - 4 | 559 | 69 | ||
AK088919.1 | - | 3 - 4 | 458 | 69 | ||
AY049978.1 | - | 3 - 4 | 480 | 69 | ||
BC022605.1 | - | 3 - 4 | 415 | 4164994 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |