Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030260.3 | NP_084536.2 | 1 - 2 | 1079 | 59 | |
NM_173002.3 | NP_766590.1 | 1 - 2 | 1079 | 59 | ||
GenBank mRNA | AK030287.1 | - | 1 - 2 | 995 | 59 | |
AK042190.1 | - | 1 - 2 | 1006 | 59 | ||
AK042392.1 | - | 1 - 2 | 989 | 59 | ||
AK044429.1 | - | 1 - 2 | 972 | 59 | ||
AK044529.1 | - | 1 - 2 | 1003 | 59 | ||
AK044948.1 | - | 1 - 2 | 973 | 59 | ||
AK047538.1 | - | 1 - 2 | 995 | 59 | ||
AK080307.1 | - | 1 - 2 | 973 | 59 | ||
AK081005.1 | - | 1 - 2 | 1081 | 59 | ||
AK141613.1 | - | 1 - 2 | 956 | 59 | ||
AK169517.1 | - | 1 - 2 | 966 | 59 | ||
BC003332.1 | - | 1 - 2 | 1063 | 3256958 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |