Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199242.1 | NP_001186171.1 | 6 - 7 | 733 | 123 | |
NM_001199243.1 | NP_001186172.1 | 5 - 6 | 631 | 123 | ||
NM_001199244.1 | NP_001186173.1 | 5 - 6 | 774 | 123 | ||
NM_001199245.1 | NP_001186174.1 | 5 - 6 | 627 | 123 | ||
NM_030265.3 | NP_084541.3 | 5 - 6 | 559 | 123 | ||
GenBank mRNA | AF305071.1 | - | 5 - 6 | 530 | 123 | |
AF453243.1 | - | 5 - 6 | 467 | 123 | ||
AK039048.1 | - | 5 - 6 | 560 | 123 | ||
AK049662.1 | - | 5 - 6 | 511 | 123 | ||
AK133416.1 | - | 5 - 6 | 479 | 123 | ||
AK148685.1 | - | 5 - 6 | 776 | 123 | ||
AY647240.1 | - | 5 - 6 | 510 | 123 | ||
BC051130.1 | - | 6 - 7 | 733 | 6532082 | 123 | |
DQ148501.1 | - | 6 - 7 | 733 | 123 | ||
DQ148502.1 | - | 4 - 5 | 491 | 123 | ||
DQ148503.1 | - | 5 - 6 | 511 | 123 | ||
DQ148504.1 | - | 5 - 6 | 627 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |