Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900014116.mm.1 |
NM_030716.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014116.mm.1 |
NM_145703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014116.mm.1 |
TC1900001590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014118.mm.1 |
NM_030716.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014118.mm.1 |
NM_145703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014118.mm.1 |
NM_145704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014118.mm.1 |
TC1900001590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014123.mm.1 |
NM_030716.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014123.mm.1 |
NM_145703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014123.mm.1 |
NM_145704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014123.mm.1 |
TC1900001590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014122.mm.1 |
NM_030716.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014122.mm.1 |
NM_145703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014122.mm.1 |
NM_145704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014122.mm.1 |
TC1900001590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014111.mm.1 |
NM_145703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014111.mm.1 |
TC1900001590.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014125.mm.1 |
NM_030716.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014125.mm.1 |
NM_145703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014125.mm.1 |
NM_145704.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014125.mm.1 |
TC1900001590.mm.1 |
MTA TC |