Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054094.5 | NP_473435.1 | 11 - 12 | 1498 | 99 | |
GenBank mRNA | AB059428.1 | - | 11 - 12 | 1502 | 99 | |
AK149550.1 | - | 11 - 12 | 1508 | 99 | ||
AK149586.1 | - | 10 - 11 | 1443 | 99 | ||
BC016414.1 | - | 11 - 12 | 1495 | 4238964 | 99 | |
BC022149.1 | - | 11 - 12 | 1476 | 5102652 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |