Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133234.2 | NP_573497.1 | 1 - 2 | 283 | 119 | |
GenBank mRNA | AF332560.1 | - | 1 - 2 | 228 | 119 | |
BC044782.2 | - | 1 - 2 | 250 | 119 | ||
BC060370.1 | - | 1 - 2 | 213 | 119 | ||
BC147261.1 | - | 1 - 2 | 111 | 9056294 | 119 | |
BC147262.1 | - | 1 - 2 | 111 | 9056313 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |