Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134791.2 | NP_001128263.1 | 12 - 13 | 1218 | 90 | |
NM_133885.4 | NP_598646.4 | 15 - 16 | 1451 | 90 | ||
NM_173350.3 | NP_775485.2 | 13 - 14 | 1257 | 90 | ||
GenBank mRNA | AK035658.1 | - | 13 - 14 | 1206 | 90 | |
AK075981.1 | - | 2 - 3 | 236 | 90 | ||
AK131984.1 | - | 15 - 16 | 90 | |||
AK172010.1 | - | 13 - 14 | 1244 | 90 | ||
BC021507.1 | - | 11 - 12 | 1079 | 5347121 | 90 | |
BC023759.1 | - | 15 - 16 | 1406 | 90 | ||
BC026927.1 | - | 13 - 14 | 1140 | 5134780 | 90 | |
BC089163.1 | - | 12 - 13 | 1159 | 5367603 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |