Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168250.1 | NP_001161722.1 | 14 - 15 | 1802 | 87 | |
NM_134092.3 | NP_598853.3 | 14 - 15 | 1802 | 87 | ||
GenBank mRNA | AJ278508.1 | - | 14 - 15 | 1723 | 87 | |
AK080036.1 | - | 14 - 15 | 1616 | 87 | ||
AK088549.1 | - | 14 - 15 | 1616 | 87 | ||
AK088652.1 | - | 4 - 5 | 1230 | 87 | ||
AK133855.1 | - | 14 - 15 | 1650 | 87 | ||
BC005500.1 | - | 7 - 8 | 799 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |