Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139153.2 | NP_631892.2 | 15 - 16 | 2074 | 144 | |
GenBank mRNA | AF459091.1 | - | 15 - 16 | 2074 | 144 | |
AK079516.1 | - | 11 - 12 | 1413 | 144 | ||
BC031173.1 | - | 6 - 7 | 646 | 144 | ||
BM943884.1 | - | 2 - 3 | 139 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |