Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139293.2 | NP_647454.2 | 8 - 9 | 1159 | 106 | |
NM_177940.1 | NP_808809.1 | 8 - 9 | 1379 | 106 | ||
NM_177941.1 | NP_808810.1 | 9 - 10 | 1466 | 106 | ||
NM_177942.1 | NP_808811.1 | 7 - 8 | 1072 | 106 | ||
GenBank mRNA | AF396699.1 | - | 8 - 9 | 1100 | 106 | |
AF489569.1 | - | 8 - 9 | 1198 | 106 | ||
AF489570.1 | - | 9 - 10 | 1285 | 106 | ||
AF489571.1 | - | 7 - 8 | 1072 | 106 | ||
AF489572.1 | - | 8 - 9 | 1159 | 106 | ||
AK031951.1 | - | 8 - 9 | 1167 | 106 | ||
AK043473.1 | - | 6 - 7 | 822 | 106 | ||
BC030900.1 | - | 3 - 4 | 282 | 106 | ||
BC138052.1 | - | 8 - 9 | 1066 | 8861071 | 106 | |
BC138053.1 | - | 8 - 9 | 1066 | 8861072 | 106 | |
BC145126.1 | - | 8 - 9 | 1066 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |