Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400002997.mm.1 |
NM_139305.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002997.mm.1 |
NM_139305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002997.mm.1 |
TC0400000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002995.mm.1 |
NM_139305.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002995.mm.1 |
NM_139305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002995.mm.1 |
TC0400000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002999.mm.1 |
NM_139305.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002999.mm.1 |
NM_139305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002999.mm.1 |
TC0400000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400002994.mm.1 |
NM_139305.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002994.mm.1 |
NM_139305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400002994.mm.1 |
TC0400000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003000.mm.1 |
NM_139305.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003000.mm.1 |
NM_139305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003000.mm.1 |
TC0400000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003001.mm.1 |
NM_139305.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003001.mm.1 |
NM_139305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003001.mm.1 |
TC0400000438.mm.1 |
MTA TC |