Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k7cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144854.2 | NP_659103.1 | 2 - 3 | 375 | 64 | |
GenBank mRNA | AK041664.1 | - | 2 - 3 | 285 | 64 | |
AY033899.1 | - | 2 - 3 | 390 | 64 | ||
BC115624.1 | - | 2 - 3 | 149 | 40102836 | 64 | |
BC115625.1 | - | 2 - 3 | 149 | 40102837 | 64 | |
BX519631.1 | - | 2 - 3 | 376 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |