Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289598.1 | NP_001276527.1 | 4 - 5 | 633 | 63 | |
NM_001289599.1 | NP_001276528.1 | 4 - 5 | 600 | 63 | ||
NM_145367.4 | NP_663342.3 | 4 - 5 | 657 | 63 | ||
GenBank mRNA | AK136046.1 | - | 4 - 5 | 545 | 63 | |
AK142049.1 | - | 4 - 5 | 643 | 63 | ||
AK150781.1 | - | 4 - 5 | 641 | 63 | ||
AK169680.1 | - | 4 - 5 | 624 | 63 | ||
AK224911.1 | - | 3 - 4 | 310 | 63 | ||
AK224913.1 | - | 4 - 5 | 644 | 63 | ||
AK224922.1 | - | 4 - 5 | 602 | 63 | ||
AK224923.1 | - | 4 - 5 | 624 | 63 | ||
AK224924.1 | - | 4 - 5 | 632 | 63 | ||
AK224926.1 | - | 4 - 5 | 602 | 63 | ||
AK224927.1 | - | 4 - 5 | 653 | 63 | ||
AK224930.1 | - | 4 - 5 | 605 | 63 | ||
AK224933.1 | - | 4 - 5 | 594 | 63 | ||
AK224934.1 | - | 4 - 5 | 648 | 63 | ||
AK224936.1 | - | 4 - 5 | 647 | 63 | ||
AK224937.1 | - | 4 - 5 | 646 | 63 | ||
AK224938.1 | - | 4 - 5 | 641 | 63 | ||
AK224939.1 | - | 4 - 5 | 634 | 63 | ||
AK224943.1 | - | 4 - 5 | 605 | 63 | ||
AK224945.1 | - | 4 - 5 | 641 | 63 | ||
AK224946.1 | - | 4 - 5 | 644 | 63 | ||
AK224947.1 | - | 4 - 5 | 631 | 63 | ||
AK224948.1 | - | 4 - 5 | 605 | 63 | ||
AK224949.1 | - | 4 - 5 | 594 | 63 | ||
AK224950.1 | - | 4 - 5 | 594 | 63 | ||
AK224951.1 | - | 4 - 5 | 652 | 63 | ||
AK224952.1 | - | 4 - 5 | 620 | 63 | ||
AK224957.1 | - | 4 - 5 | 610 | 63 | ||
AK224958.1 | - | 4 - 5 | 610 | 63 | ||
AK224960.1 | - | 4 - 5 | 652 | 63 | ||
AK224962.1 | - | 4 - 5 | 612 | 63 | ||
AK224963.1 | - | 4 - 5 | 653 | 63 | ||
AK224964.1 | - | 4 - 5 | 653 | 63 | ||
AK224965.1 | - | 4 - 5 | 629 | 63 | ||
AK224966.1 | - | 4 - 5 | 629 | 63 | ||
AK224968.1 | - | 2 - 3 | 187 | 63 | ||
AK224969.1 | - | 2 - 3 | 187 | 63 | ||
AK224970.1 | - | 4 - 5 | 594 | 63 | ||
AK224971.1 | - | 4 - 5 | 654 | 63 | ||
AK224972.1 | - | 4 - 5 | 644 | 63 | ||
AK224973.1 | - | 4 - 5 | 594 | 63 | ||
AK224974.1 | - | 4 - 5 | 597 | 63 | ||
AK224976.1 | - | 4 - 5 | 627 | 63 | ||
AK224977.1 | - | 4 - 5 | 641 | 63 | ||
AK224978.1 | - | 4 - 5 | 641 | 63 | ||
AK224979.1 | - | 4 - 5 | 628 | 63 | ||
AK224981.1 | - | 4 - 5 | 598 | 63 | ||
AK224982.1 | - | 4 - 5 | 653 | 63 | ||
AK224983.1 | - | 4 - 5 | 642 | 63 | ||
AK224984.1 | - | 4 - 5 | 607 | 63 | ||
AK224985.1 | - | 4 - 5 | 611 | 63 | ||
AK224986.1 | - | 4 - 5 | 655 | 63 | ||
AK224988.1 | - | 4 - 5 | 631 | 63 | ||
AK224989.1 | - | 4 - 5 | 648 | 63 | ||
AK224990.1 | - | 4 - 5 | 643 | 63 | ||
AK224991.1 | - | 4 - 5 | 638 | 63 | ||
AK224992.1 | - | 4 - 5 | 638 | 63 | ||
AK224993.1 | - | 4 - 5 | 638 | 63 | ||
AK224994.1 | - | 4 - 5 | 646 | 63 | ||
AK224995.1 | - | 4 - 5 | 609 | 63 | ||
AK224996.1 | - | 4 - 5 | 580 | 63 | ||
AK224997.1 | - | 4 - 5 | 605 | 63 | ||
AK224998.1 | - | 4 - 5 | 603 | 63 | ||
AK224999.1 | - | 4 - 5 | 625 | 63 | ||
AK225004.1 | - | 4 - 5 | 646 | 63 | ||
AK225005.1 | - | 4 - 5 | 624 | 63 | ||
AK225007.1 | - | 4 - 5 | 646 | 63 | ||
AK225010.1 | - | 4 - 5 | 646 | 63 | ||
AK225011.1 | - | 4 - 5 | 646 | 63 | ||
AK225012.1 | - | 4 - 5 | 415 | 63 | ||
AK225013.1 | - | 4 - 5 | 415 | 63 | ||
AK225014.1 | - | 4 - 5 | 624 | 63 | ||
AK225015.1 | - | 4 - 5 | 415 | 63 | ||
AK225016.1 | - | 4 - 5 | 554 | 63 | ||
AK225017.1 | - | 4 - 5 | 602 | 63 | ||
AK225019.1 | - | 4 - 5 | 634 | 63 | ||
AK225020.1 | - | 4 - 5 | 585 | 63 | ||
AK225021.1 | - | 4 - 5 | 585 | 63 | ||
AK225022.1 | - | 4 - 5 | 433 | 63 | ||
AK225023.1 | - | 4 - 5 | 659 | 63 | ||
AY243534.1 | - | 4 - 5 | 620 | 63 | ||
AY548112.1 | - | 4 - 5 | 581 | 63 | ||
BC016252.1 | - | 4 - 5 | 624 | 4911455 | 63 | |
BC024505.1 | - | 4 - 5 | 569 | 4983496 | 63 | |
BC036155.1 | - | 3 - 4 | 243 | 63 | ||
BC046789.1 | - | 4 - 5 | 583 | 5716897 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |