Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289598.1 | NP_001276527.1 | 8 - 9 | 1051 | 89 | |
NM_001289599.1 | NP_001276528.1 | 8 - 9 | 1018 | 89 | ||
NM_145367.4 | NP_663342.3 | 8 - 9 | 1075 | 89 | ||
GenBank mRNA | AK136046.1 | - | 8 - 9 | 963 | 89 | |
AK142049.1 | - | 8 - 9 | 1061 | 89 | ||
AK150781.1 | - | 8 - 9 | 1059 | 89 | ||
AK169680.1 | - | 8 - 9 | 1042 | 89 | ||
AK224911.1 | - | 7 - 8 | 728 | 89 | ||
AK224913.1 | - | 8 - 9 | 1062 | 89 | ||
AK224922.1 | - | 8 - 9 | 1020 | 89 | ||
AK224923.1 | - | 8 - 9 | 1042 | 89 | ||
AK224924.1 | - | 8 - 9 | 1050 | 89 | ||
AK224926.1 | - | 8 - 9 | 1020 | 89 | ||
AK224927.1 | - | 8 - 9 | 1071 | 89 | ||
AK224930.1 | - | 8 - 9 | 1023 | 89 | ||
AK224933.1 | - | 8 - 9 | 1012 | 89 | ||
AK224934.1 | - | 8 - 9 | 1066 | 89 | ||
AK224935.1 | - | 1 - 2 | 1217 | 89 | ||
AK224936.1 | - | 8 - 9 | 1065 | 89 | ||
AK224937.1 | - | 8 - 9 | 1064 | 89 | ||
AK224938.1 | - | 8 - 9 | 1059 | 89 | ||
AK224939.1 | - | 8 - 9 | 1052 | 89 | ||
AK224943.1 | - | 8 - 9 | 1023 | 89 | ||
AK224945.1 | - | 8 - 9 | 1059 | 89 | ||
AK224946.1 | - | 8 - 9 | 1062 | 89 | ||
AK224947.1 | - | 8 - 9 | 1049 | 89 | ||
AK224948.1 | - | 8 - 9 | 1023 | 89 | ||
AK224949.1 | - | 8 - 9 | 1012 | 89 | ||
AK224950.1 | - | 8 - 9 | 1012 | 89 | ||
AK224951.1 | - | 8 - 9 | 1070 | 89 | ||
AK224955.1 | - | 3 - 4 | 323 | 89 | ||
AK224956.1 | - | 8 - 9 | 982 | 89 | ||
AK224957.1 | - | 8 - 9 | 1028 | 89 | ||
AK224958.1 | - | 8 - 9 | 1028 | 89 | ||
AK224960.1 | - | 8 - 9 | 1070 | 89 | ||
AK224962.1 | - | 8 - 9 | 1030 | 89 | ||
AK224965.1 | - | 8 - 9 | 1047 | 89 | ||
AK224966.1 | - | 8 - 9 | 1047 | 89 | ||
AK224967.1 | - | 8 - 9 | 1038 | 89 | ||
AK224968.1 | - | 6 - 7 | 605 | 89 | ||
AK224969.1 | - | 6 - 7 | 605 | 89 | ||
AK224970.1 | - | 8 - 9 | 1012 | 89 | ||
AK224971.1 | - | 8 - 9 | 1072 | 89 | ||
AK224972.1 | - | 8 - 9 | 1062 | 89 | ||
AK224973.1 | - | 8 - 9 | 1012 | 89 | ||
AK224974.1 | - | 8 - 9 | 1015 | 89 | ||
AK224975.1 | - | 3 - 4 | 250 | 89 | ||
AK224976.1 | - | 8 - 9 | 1045 | 89 | ||
AK224977.1 | - | 8 - 9 | 1059 | 89 | ||
AK224978.1 | - | 8 - 9 | 1059 | 89 | ||
AK224979.1 | - | 8 - 9 | 1046 | 89 | ||
AK224981.1 | - | 8 - 9 | 1016 | 89 | ||
AK224982.1 | - | 8 - 9 | 1071 | 89 | ||
AK224983.1 | - | 8 - 9 | 1060 | 89 | ||
AK224984.1 | - | 8 - 9 | 1025 | 89 | ||
AK224986.1 | - | 8 - 9 | 1073 | 89 | ||
AK224988.1 | - | 8 - 9 | 1049 | 89 | ||
AK224989.1 | - | 8 - 9 | 1066 | 89 | ||
AK224990.1 | - | 8 - 9 | 1061 | 89 | ||
AK224991.1 | - | 8 - 9 | 1056 | 89 | ||
AK224992.1 | - | 8 - 9 | 1056 | 89 | ||
AK224993.1 | - | 8 - 9 | 1056 | 89 | ||
AK224994.1 | - | 8 - 9 | 1064 | 89 | ||
AK224995.1 | - | 8 - 9 | 1027 | 89 | ||
AK224996.1 | - | 8 - 9 | 998 | 89 | ||
AK224997.1 | - | 8 - 9 | 1023 | 89 | ||
AK224998.1 | - | 8 - 9 | 1021 | 89 | ||
AK224999.1 | - | 8 - 9 | 1043 | 89 | ||
AK225004.1 | - | 8 - 9 | 1064 | 89 | ||
AK225005.1 | - | 8 - 9 | 1042 | 89 | ||
AK225007.1 | - | 8 - 9 | 1064 | 89 | ||
AK225010.1 | - | 8 - 9 | 1064 | 89 | ||
AK225011.1 | - | 8 - 9 | 1064 | 89 | ||
AK225012.1 | - | 8 - 9 | 833 | 89 | ||
AK225013.1 | - | 8 - 9 | 833 | 89 | ||
AK225014.1 | - | 8 - 9 | 1042 | 89 | ||
AK225015.1 | - | 8 - 9 | 833 | 89 | ||
AK225016.1 | - | 8 - 9 | 972 | 89 | ||
AK225017.1 | - | 8 - 9 | 1020 | 89 | ||
AK225019.1 | - | 8 - 9 | 1052 | 89 | ||
AK225020.1 | - | 8 - 9 | 1003 | 89 | ||
AK225021.1 | - | 8 - 9 | 1003 | 89 | ||
AK225022.1 | - | 8 - 9 | 851 | 89 | ||
AK225023.1 | - | 8 - 9 | 1077 | 89 | ||
AY243534.1 | - | 8 - 9 | 1038 | 89 | ||
AY548112.1 | - | 8 - 9 | 999 | 89 | ||
BC016252.1 | - | 8 - 9 | 1042 | 4911455 | 89 | |
BC024505.1 | - | 8 - 9 | 987 | 4983496 | 89 | |
BC036155.1 | - | 7 - 8 | 661 | 89 | ||
BC046789.1 | - | 8 - 9 | 1001 | 5716897 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |