Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181328.3 | NP_851845.1 | 1 - 2 | 226 | 56 | |
GenBank mRNA | AK047628.1 | - | 1 - 2 | 227 | 56 | |
AK087057.1 | - | 1 - 2 | 213 | 56 | ||
AK148350.1 | - | 1 - 2 | 204 | 56 | ||
AK160906.1 | - | 1 - 2 | 175 | 56 | ||
BC006711.1 | - | 1 - 2 | 99 | 3584570 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |