Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100025274.mm.1 |
NM_001159624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025274.mm.1 |
NM_001159624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025274.mm.1 |
NM_145417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025274.mm.1 |
NM_145417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025274.mm.1 |
TC0100003124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025272.mm.1 |
NM_001159624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025272.mm.1 |
NM_001159624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025272.mm.1 |
NM_145417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025272.mm.1 |
NM_145417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025272.mm.1 |
TC0100003124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025277.mm.1 |
NM_001159624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025277.mm.1 |
NM_001159624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025277.mm.1 |
NM_145417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025277.mm.1 |
NM_145417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025277.mm.1 |
TC0100003124.mm.1 |
MTA TC |