Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145431.2 | NP_663406.2 | 4 - 5 | 501 | 82 | |
GenBank mRNA | AK141230.1 | - | 4 - 5 | 476 | 82 | |
AK170853.1 | - | 4 - 5 | 474 | 82 | ||
BC018399.1 | - | 4 - 5 | 492 | 3491925 | 82 | |
BC035421.1 | - | 4 - 5 | 492 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |