Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252532.1 | NP_001239461.1 | 15 - 16 | 2027 | 74 | |
NM_145438.2 | NP_663413.2 | 15 - 16 | 2027 | 74 | ||
GenBank mRNA | AK143486.1 | - | 15 - 16 | 2015 | 74 | |
AK143798.1 | - | 15 - 16 | 2002 | 74 | ||
AK167536.1 | - | 15 - 16 | 1999 | 74 | ||
AY033650.1 | - | 15 - 16 | 1977 | 74 | ||
BC013232.1 | - | 11 - 12 | 1568 | 74 | ||
BC037226.1 | - | 15 - 16 | 1999 | 3584689 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |