Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145494.2 | NP_663469.1 | 12 - 13 | 1411 | 76 | |
GenBank mRNA | AK042289.1 | - | 12 - 13 | 1458 | 76 | |
AK043103.1 | - | 12 - 13 | 1448 | 76 | ||
AK050933.1 | - | 12 - 13 | 1458 | 76 | ||
AK050980.1 | - | 12 - 13 | 1458 | 76 | ||
AK077746.1 | - | 12 - 13 | 1458 | 76 | ||
AK156403.1 | - | 12 - 13 | 1458 | 76 | ||
AK162126.1 | - | 12 - 13 | 1470 | 76 | ||
AK171157.1 | - | 12 - 13 | 1458 | 76 | ||
BC004709.1 | - | 12 - 13 | 1411 | 3499365 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |