Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164359.1 | NP_001157831.1 | 8 - 9 | 763 | 76 | |
NM_001164360.1 | NP_001157832.1 | 8 - 9 | 671 | 76 | ||
NM_145502.3 | NP_663477.3 | 7 - 8 | 867 | 76 | ||
GenBank mRNA | AK133557.1 | - | 7 - 8 | 610 | 76 | |
AK145990.1 | - | 8 - 9 | 644 | 76 | ||
BC011220.1 | - | 8 - 9 | 635 | 4208238 | 76 | |
BC023849.1 | - | 7 - 8 | 730 | 5345801 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |