Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145594.2 | NP_663569.2 | 1 - 2 | 76 | 80 | |
GenBank mRNA | AF478470.1 | - | 1 - 2 | 86 | 80 | |
AK083397.1 | - | 1 - 2 | 74 | 80 | ||
BC021946.1 | - | 1 - 2 | 44 | 5098805 | 80 | |
BC029734.1 | - | 1 - 2 | 53 | 4158843 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |