Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145621.2 | NP_663596.1 | 9 - 10 | 991 | 79 | |
GenBank mRNA | AK085143.1 | - | 998 | 79 | ||
AK147323.1 | - | 9 - 10 | 979 | 79 | ||
BC017634.1 | - | 9 - 10 | 991 | 4527857 | 79 | |
BC022169.1 | - | 7 - 8 | 1357 | 79 | ||
BC086762.1 | - | 8 - 9 | 1189 | 79 | ||
BC111033.1 | - | 9 - 10 | 986 | 5368225 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |