Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043344.1 | - | 3 - 4 | 435 | 112 | |
AK145200.1 | - | 4 - 5 | 491 | 112 | ||
AK161473.1 | - | 4 - 5 | 574 | 112 | ||
BC030328.1 | - | 4 - 5 | 491 | 5376020 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |