Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145934.2 | NP_666046.1 | 7 - 8 | 738 | 101 | |
NR_102313.1 | 7 - 8 | 738 | 101 | |||
GenBank mRNA | AK050131.1 | - | 7 - 8 | 718 | 101 | |
AK050172.1 | - | 7 - 8 | 706 | 101 | ||
BC026642.1 | - | 7 - 8 | 712 | 4986113 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |