Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008498.2 | NP_032524.1 | 2 - 3 | 1758 | 68 | |
GenBank mRNA | BC092374.1 | - | 2 - 3 | 1891 | 30363447 | 68 |
S68107.1 | - | 5 - 6 | 1290 | 68 | ||
Z27410.1 | - | 2 - 3 | 1645 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |