Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lta4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008517.2 | NP_032543.2 | 4 - 5 | 71 | ||
GenBank mRNA | AK134931.1 | - | 4 - 5 | 71 | ||
BC021417.1 | - | 4 - 5 | 588 | 5004113 | 71 | |
M63848.1 | - | 560 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |